siRNA Details
Gene Symbol: Dync1li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146229.2 | NP_666341.1 | 6 | 821 | ||
GenBank mRNA | AK159702.1 | - | 863 | |||
AK183051.1 | - | 2 | 181 | |||
AK189933.1 | - | 2 | 180 | |||
AK191001.1 | - | 2 | 180 | |||
AK191112.1 | - | 2 | 182 | |||
AK191959.1 | - | 2 | 180 | |||
AK193282.1 | - | 2 | 180 | |||
AK194365.1 | - | 2 | 180 | |||
AK195148.1 | - | 2 | 180 | |||
AK195418.1 | - | 2 | 180 | |||
AK196420.1 | - | 2 | 181 | |||
AK196819.1 | - | 2 | 181 | |||
AK197477.1 | - | 2 | 180 | |||
AK197747.1 | - | 2 | 180 | |||
AK198065.1 | - | 2 | 180 | |||
AK201083.1 | - | 220 | ||||
AK202293.1 | - | 2 | 180 | |||
AK202548.1 | - | 2 | 179 | |||
AK204826.1 | - | 2 | 180 | |||
AK213717.1 | - | 2 | 180 | |||
AK218794.1 | - | 2 | 180 | |||
BC023347.1 | - | 6 | 821 | 5040044 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
