siRNA Details
Gene Symbol: Dync1li1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146229.2 | NP_666341.1 | 8 | 1036 | ||
GenBank mRNA | AK159702.1 | - | 1078 | |||
AK159983.1 | - | 25 | ||||
AK205150.1 | - | 2 | 97 | |||
AK217382.1 | - | 2 | 48 | |||
BC023347.1 | - | 8 | 1036 | 5040044 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_145772, AF181992.
Gene Ontology Categories:
