siRNA Details
Gene Symbol: Dhrsx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033326.2 | NP_001028498.2 | 7 | 2112 | ||
GenBank mRNA | AK030649.1 | - | 6 | 2040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
