siRNA Details
Gene Symbol: Pyhin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175026.3 | NP_778191.1 | 6 | 1529 | ||
GenBank mRNA | AK076726.1 | - | 2 | 407 | ||
AK080371.1 | - | 6 | 1519 | |||
AK150817.1 | - | 6 | 1420 | |||
BC096384.1 | - | 6 | 1483 | 30752094 | ||
BC107378.1 | - | 7 | 1471 | 40056703 | ||
BC107379.1 | - | 6 | 1363 | 40056704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
