siRNA Details
Gene Symbol: 5730507C01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033157.4 | NP_001028329.1 | 4 | 669 | ||
NM_001201330.1 | NP_001188259.1 | 4 | 741 | |||
GenBank mRNA | AK017753.1 | - | 4 | 741 | ||
AK139861.1 | - | 4 | 635 | |||
BC150925.1 | - | 4 | 532 | 9087836 | ||
BY717547.1 | - | 4 | 741 |
