siRNA Details
Gene Symbol: Gm4907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034864.3 | NP_001030036.2 | 2 | 1118 | ||
GenBank mRNA | AK133369.1 | - | 1245 | |||
BC147618.1 | - | 2 | 1118 | 9056608 | ||
BC147627.1 | - | 2 | 1118 | 9056617 |
