siRNA Details
Gene Symbol: Gpr119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181751.2 | NP_861416.1 | 1 | 891 | ||
GenBank mRNA | AB233291.1 | - | 1 | 891 | ||
AY288423.1 | - | 1 | 671 | |||
BC115557.1 | - | 1 | 671 | 40101841 | ||
BC115558.1 | - | 1 | 671 | 40101842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
