siRNA Details
Gene Symbol: Gemin8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146238.3 | NP_666350.2 | 4 | 773 | ||
GenBank mRNA | AK037580.1 | - | 4 | 670 | ||
AK047511.1 | - | 5 | 868 | |||
AK053774.1 | - | 4 | 737 | |||
AK160583.1 | - | 4 | 774 | |||
BC023488.1 | - | 5 | 763 | 5010787 |

More Information
Other Species Targeted

Gene Ontology Categories:
