siRNA Details
Gene Symbol: Ofd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177429.3 | NP_803178.2 | 16 | 2084 | ||
GenBank mRNA | AJ438159.1 | - | 16 | 1935 | ||
BC119070.1 | - | 16 | 1767 | 8733819 | ||
BC120487.1 | - | 16 | 1767 | 40129410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
