siRNA Details
Gene Symbol: Tbpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011603.5 | NP_035733.1 | 6 | 812 | ||
GenBank mRNA | AB017697.1 | - | 6 | 709 | ||
AK005604.1 | - | 6 | 631 | |||
AK049975.1 | - | 6 | 684 | |||
AK078547.1 | - | 6 | 744 | |||
BC033926.1 | - | 6 | 653 | 1382180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
