siRNA Details
Gene Symbol: Rpl27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011975.3 | NP_036105.2 | 4 | 248 | ||
GenBank mRNA | AK007837.1 | - | 4 | 247 | ||
AK008531.1 | - | 4 | 246 | |||
AK010286.1 | - | 4 | 249 | |||
AK011361.1 | - | 4 | 248 | |||
AK138110.1 | - | 2 | 991 | |||
AK150907.1 | - | 4 | 246 | |||
AK167792.1 | - | 4 | 247 | |||
AK167915.1 | - | 4 | 247 | |||
BC056958.1 | - | 3 | 232 | 6817520 | ||
BC081430.1 | - | 3 | 232 | 6411497 | ||
BC086939.1 | - | 4 | 245 | 6760797 | ||
CJ072009.1 | - | 4 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
