siRNA Details
Gene Symbol: Gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033331.2 | NP_001028503.2 | 11 | 2017 | ||
NM_001079876.1 | NP_001073345.1 | 11 | 2020 | |||
GenBank mRNA | AK136054.1 | - | 11 | 2030 | ||
AK170634.1 | - | 11 | 1952 | |||
AK170750.1 | - | 11 | 1942 |
