siRNA Details
Gene Symbol: Pla2g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172791.2 | NP_766379.2 | 2 | 697 | ||
GenBank mRNA | AK033572.1 | - | 2 | 641 | ||
AK046215.1 | - | 1 | 842 | |||
AK081075.1 | - | 2 | 625 | |||
BC079556.1 | - | 2 | 697 | 30360278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
