siRNA Details
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172793.2 | NP_766381.1 | 3 | 296 | ||
GenBank mRNA | AK080918.1 | - | 3 | 279 | ||
AK084528.1 | - | 3 | 297 | |||
AY177687.1 | - | 3 | 263 | |||
BC025540.1 | - | 3 | 274 | |||
BC052556.1 | - | 2 | 209 | |||
BC150808.1 | - | 3 | 253 | 9087719 | ||
BC151155.1 | - | 3 | 253 | 9088043 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
