siRNA Details
Gene Symbol: Mief2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009927.2 | NP_001009927.1 | 4 | 508 | ||
GenBank mRNA | AK156917.1 | - | 4 | 491 | ||
AK171444.1 | - | 3 | 633 | |||
AK211527.1 | - | 3 | 229 | |||
BC088984.1 | - | 4 | 397 | 5708020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
