siRNA Details
Gene Symbol: 1700071K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033765.2 | NP_001028937.1 | 1 | 291 | ||
GenBank mRNA | AK131752.1 | - | 1 | 292 | ||
BC139179.1 | - | 1 | 257 | 8862201 | ||
BC139181.1 | - | 1 | 257 | 8862203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
