siRNA Details
Gene Symbol: Slfn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172796.2 | NP_766384.2 | 5 | 2622 | ||
GenBank mRNA | AK036579.1 | - | 5 | 2623 | ||
AY217035.1 | - | 4 | 2090 | |||
AY261800.1 | - | 5 | 2530 | |||
AY261801.1 | - | 5 | 2512 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
