siRNA Details
Gene Symbol: Slfn10-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144698.1 | - | 2 | 289 | ||
AK220241.1 | - | 2 | 413 | |||
AY261802.1 | - | 2 | 288 | |||
AY261803.1 | - | 2 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
