siRNA Details
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 13 | 1263 | ||
GenBank mRNA | AK050594.1 | - | 12 | 1070 | ||
AK149171.1 | - | 3 | 323 | |||
AY116674.1 | - | 13 | 1149 | |||
BC025436.1 | - | 13 | 1251 | |||
BC058209.1 | - | 13 | 1180 | |||
BC080780.1 | - | 13 | 1198 | |||
BC157975.1 | - | 12 | 1065 | 9007297 | ||
BC158109.1 | - | 12 | 1065 | 9007390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
