siRNA Details
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 20 | 3359 | ||
GenBank mRNA | BC037715.1 | - | 4 | 1350 | ||
BC157975.1 | - | 19 | 3158 | 9007297 | ||
BC158109.1 | - | 19 | 3161 | 9007390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
