siRNA Details
Gene Symbol: Kcnh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172805.3 | NP_766393.2 | 8 | 2264 | ||
GenBank mRNA | AK032438.1 | - | 8 | 2265 | ||
BC066138.1 | - | 8 | 1606 | 30094199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
