siRNA Details
Gene Symbol: Zfp458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 1326 | ||
GenBank mRNA | AB010368.1 | - | 1 | 225 | ||
AK133308.1 | - | 1244 | ||||
BC062958.1 | - | 4 | 1326 | 30548236 |

More Information
Other Species Targeted

Gene Ontology Categories:
