siRNA Details
Gene Symbol: Gpr150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175495.2 | NP_780704.1 | 1 | 224 | ||
GenBank mRNA | AK047609.1 | - | 1 | 224 | ||
AY255541.1 | - | 1 | 28 | |||
BC118612.1 | - | 1 | 204 | 40107174 | ||
BC118614.1 | - | 1 | 205 | 40107172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
