siRNA Details
Gene Symbol: Tnpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048267.1 | NP_001041732.1 | 23 | 2755 | ||
NM_178716.3 | NP_848831.2 | 23 | 2761 | |||
GenBank mRNA | AK029953.1 | - | 23 | 2762 | ||
AK030580.1 | - | 23 | 2721 | |||
AK045132.1 | - | 23 | 2755 | |||
AK163279.1 | - | 6 | 424 | |||
BC031571.1 | - | 15 | 1804 | |||
BC055372.1 | - | 23 | 2662 | 30059046 |

More Information
Other Species Targeted

Gene Ontology Categories:
