siRNA Details
Gene Symbol: Lrit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173418.3 | NP_775594.1 | 2 | 478 | ||
GenBank mRNA | AK044383.1 | - | 2 | 479 | ||
AK044611.1 | - | 2 | 480 | |||
BC037728.1 | - | 2 | 378 | |||
BC057628.1 | - | 2 | 453 | 4511498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
