siRNA Details
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199470.2 | NP_955764.1 | 2 | 418 | ||
GenBank mRNA | BC057373.1 | - | 372 | 6817232 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770708.
Gene Ontology Categories:
