siRNA Details
Gene Symbol: Il17d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145837.3 | NP_665836.2 | 3 | 913 | ||
GenBank mRNA | AF458063.1 | - | 2 | 835 | ||
AK131917.1 | - | 3 | 962 | |||
BC099514.1 | - | 1 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
