siRNA Details
Gene Symbol: Dleu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173419.2 | NP_775595.1 | 2 | 465 | ||
GenBank mRNA | AK082171.1 | - | 3 | 227 | ||
BC038059.1 | - | 2 | 458 | 4235022 | ||
CK618986.1 | - | 2 | 465 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
