siRNA Details
Gene Symbol: Oxgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001490.3 | NP_001001490.1 | 4 | 1472 | ||
GenBank mRNA | AK142510.1 | - | 4 | 1473 | ||
AY612852.1 | - | 1 | 949 | |||
BC119196.1 | - | 1 | 1002 | 8733945 | ||
BC119200.1 | - | 1 | 1002 | 8733949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
