siRNA Details
Gene Symbol: Erich5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173421.2 | NP_775597.1 | 2 | 423 | ||
GenBank mRNA | AK135792.1 | - | 2 | 154 | ||
AK162262.1 | - | 2 | 181 | |||
BC030476.1 | - | 2 | 431 | 5368170 | ||
BC099414.1 | - | 2 | 174 | 30931867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
