siRNA Details
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172814.3 | NP_766402.1 | 5 | 953 | ||
GenBank mRNA | AK050512.1 | - | 5 | 954 | ||
BC058345.1 | - | 4 | 1133 | 6822657 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
