siRNA Details
Gene Symbol: Rspo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172815.3 | NP_766403.1 | 2, 3 | 976 | ||
GenBank mRNA | AK049891.1 | - | 2, 3 | 727 | ||
AK052457.1 | - | 1, 2 | 1100 | |||
AK087485.1 | - | 2, 3 | 977 | |||
BC052844.1 | - | 2, 3 | 327 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001130575, XM_008765455.
Gene Ontology Categories:
