siRNA Details
Gene Symbol: Aard | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175503.3 | NP_780712.2 | 2 | 462 | ||
GenBank mRNA | AK046414.1 | - | 2 | 463 | ||
AY134665.1 | - | 2 | 401 | |||
BC089505.1 | - | 2 | 416 | 6772439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
