siRNA Details
Gene Symbol: Colec10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173422.3 | NP_775598.2 | 5, 6 | 444 | ||
GenBank mRNA | AB016429.1 | - | 5, 6 | 448 | ||
AK028423.1 | - | 5, 6 | 446 | |||
BC106836.1 | - | 5, 6 | 446 | 40044882 | ||
BC106837.1 | - | 5, 6 | 446 | 40044883 | ||
BC108945.1 | - | 5, 6 | 446 | 40043083 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
