siRNA Details
Gene Symbol: Gpr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173365.2 | NP_775541.1 | 2 | 597 | ||
GenBank mRNA | AK040547.1 | - | 2 | 598 | ||
AY255575.1 | - | 1 | 142 | |||
BC117783.2 | - | 2 | 459 | 40092101 |

More Information
Other Species Targeted

Gene Ontology Categories:
