siRNA Details
Gene Symbol: Dip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159361.1 | NP_001152833.1 | 24 | 3028 | ||
NM_172819.3 | NP_766407.2 | 19 | 2240 | |||
GenBank mRNA | AB093297.1 | - | 9 | 1044 | ||
AK030064.1 | - | 19 | 2241 | |||
AK078824.1 | - | 9 | 1068 | |||
AK090139.1 | - | 4 | 450 | |||
AK147562.1 | - | 24 | 2964 | |||
BC023757.1 | - | 10 | 1127 | |||
BC137857.1 | - | 19 | 2227 | 8860876 | ||
BC137858.1 | - | 19 | 2227 | 8860877 |
