siRNA Details
Gene Symbol: Dip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159361.1 | NP_001152833.1 | 16 | 2050 | ||
NM_172819.3 | NP_766407.2 | 11 | 1262 | |||
GenBank mRNA | AB093297.1 | - | 1 | 66 | ||
AK030064.1 | - | 11 | 1263 | |||
AK078824.1 | - | 1 | 86 | |||
AK147562.1 | - | 16 | 1986 | |||
BC023757.1 | - | 2 | 149 | |||
BC137857.1 | - | 11 | 1249 | 8860876 | ||
BC137858.1 | - | 11 | 1249 | 8860877 |
