siRNA Details
Gene Symbol: 4732456N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177717.4 | NP_808385.2 | 2 | 687 | ||
GenBank mRNA | AK028791.1 | - | 2 | 688 | ||
BC118024.1 | - | 2 | 646 | 40092535 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4732456N10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177717.4 | NP_808385.2 | 2 | 687 | ||
GenBank mRNA | AK028791.1 | - | 2 | 688 | ||
BC118024.1 | - | 2 | 646 | 40092535 |