siRNA Details
Gene Symbol: Gmnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013761.2 | NP_001013783.1 | 6 | 913 | ||
NM_001285916.1 | NP_001272845.1 | 5 | 1007 | |||
NM_001285918.1 | NP_001272847.1 | 5 | 738 | |||
GenBank mRNA | AK141059.1 | - | 5 | 1007 | ||
AK160719.1 | - | 5 | 707 | |||
BC086669.1 | - | 5 | 668 | 30639759 |
