siRNA Details
Gene Symbol: Ostn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198112.2 | NP_932780.1 | 3 | 346 | ||
GenBank mRNA | AY395730.1 | - | 3 | 346 | ||
AY573932.1 | - | 3 | 498 | |||
BC117063.1 | - | 3 | 334 | 40126382 | ||
BC117089.1 | - | 3 | 334 | 40126408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
