siRNA Details
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172824.3 | NP_766412.2 | 6 | 607 | ||
GenBank mRNA | AK029725.1 | - | 6 | 608 | ||
AK090294.1 | - | 6 | 514 | |||
AK172014.1 | - | 1 | 861 | |||
BB618371.1 | - | 6 | 509 | |||
BC031204.1 | - | 6 | 503 | 5066612 | ||
BC095999.1 | - | 6 | 503 | |||
CF742918.1 | - | 4 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
