siRNA Details
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172824.3 | NP_766412.2 | 6 | 556 | ||
GenBank mRNA | AK029725.1 | - | 6 | 557 | ||
AK090294.1 | - | 6 | 463 | |||
AK172014.1 | - | 1 | 810 | |||
BB615458.1 | - | 6 | 557 | |||
BB618371.1 | - | 6 | 458 | |||
BC031204.1 | - | 6 | 452 | 5066612 | ||
BC095999.1 | - | 6 | 452 | |||
CF742918.1 | - | 4 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
