siRNA Details
Gene Symbol: Gpr156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153394.2 | NP_700443.2 | 10 | 2314 | ||
GenBank mRNA | AF488740.1 | - | 9 | 1838 | ||
AK141990.1 | - | 10 | 2360 | |||
BC059227.1 | - | 10 | 2314 | 6400663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
