siRNA Details
Gene Symbol: Cd200r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207244.2 | NP_997127.1 | 2 | 269 | ||
GenBank mRNA | AK089262.1 | - | 4 | 564 | ||
AY230200.1 | - | 2 | 269 | |||
BC131946.1 | - | 2 | 259 | 40130223 | ||
BC131972.1 | - | 2 | 259 | 40130249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
