siRNA Details
Gene Symbol: Arid1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085355.1 | NP_001078824.1 | 2 | 1464 | ||
GenBank mRNA | AK160334.1 | - | 1 | 190 | ||
BF462904.1 | - | 2 | 117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
