siRNA Details
Gene Symbol: St6gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172829.2 | NP_766417.1 | 3 | 1144 | ||
GenBank mRNA | AK082566.1 | - | 3 | 1145 | ||
AK129462.1 | - | 1 | 76 | |||
BC125586.1 | - | 2 | 1014 | 40130101 | ||
BC138142.1 | - | 2 | 1014 | 8861161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
