siRNA Details
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166493.1 | NP_001159965.1 | 5 | 714 | ||
NM_207246.4 | NP_997129.1 | 6 | 706 | |||
GenBank mRNA | AB154824.1 | - | 4 | 401 | ||
AK173041.1 | - | 5 | 599 | |||
BC066069.1 | - | 4 | 344 | 6851971 | ||
BC066127.1 | - | 6 | 631 | 30280332 | ||
CD348455.1 | - | 4 | 354 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108009, XM_006239759, XM_006239758, XM_006239760, XM_006239761, and others.
Gene Ontology Categories:
