siRNA Details
Gene Symbol: 9430020K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081963.1 | NP_001075432.1 | 3 | 2636 | ||
GenBank mRNA | AK220394.1 | - | 1 | 793 | ||
BC144919.1 | - | 2 | 2540 | 9053467 |
