siRNA Details
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081371.2 | NP_001074840.2 | 24 | 5640 | ||
GenBank mRNA | AY590892.1 | - | 24 | 5640 |

More Information
Other Species Targeted

Gene Ontology Categories:
