siRNA Details
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172835.3 | NP_766423.3 | 2, 3 | 304 | ||
GenBank mRNA | AK044418.1 | - | 2, 3 | 251 | ||
AK135575.1 | - | 2, 3 | 253 | |||
BC028931.1 | - | 2, 3 | 262 | 5037308 | ||
BY128359.1 | - | 304 |

More Information
Other Species Targeted

Gene Ontology Categories:
